MES, monohydrate, GMP, Excipient Grade
MES, monohydrate, GMP, Excipient Grade
BioSpectra

MES, monohydrate, GMP, Excipient Grade

This item is suitable for use as an excipient, and is manufactured in accordance with the ICH-Q7 Good Manufacturing PracticeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
BMESM-3220-1010 kg
BMESM-3220-2525 kg
BMESM-3220-5050 kg
Catalog number BMESM-3220-10
Price (INR)
-
Request A Quote
Quantity:
10 kg
This item is suitable for use as an excipient, and is manufactured in accordance with the ICH-Q7 Good Manufacturing Practice Guide. This grade of MES, monohydrate is not suitable to be used as an active pharmaceutical ingredient, drug product, or household item.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS145224-94-8
Chemical Name or MaterialMES monohydrate
ColorWhite
Molecular FormulaC6H13NO4S·H2O
PackagingHDPE round pail with lid, LDPE liner
Synonym4-Morpholineethanesulfonic Acid Monohydrate, 2-(N-Morpholino) Ethane Sulfonic Acid
Molecular Weight (g/mol)213.25
Percent Purity≥99.0%
Physical FormCrystals
Quantity10 kg
pH3.1–3.5 (5% solution)
Unit Size10 kg