Support Pack Standard
Invitrogen™

Support Pack Standard

10 x 96 tests
Have Questions?
Catalog NumberQuantity
BMS41210 x 96 Tests
Catalog number BMS412
Price (INR)
21,958.00
Online offer
Ends: 31-Dec-2026
36,595.02
Save 14,637.02 (40%)
Each
Quantity:
10 x 96 Tests
Price (INR)
21,958.00
Online offer
Ends: 31-Dec-2026
36,595.02
Save 14,637.02 (40%)
Each
Buffer pack for 10 plates

Reported Application
ELISA

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialAssay Buffer, Wash Buffer
For Use With (Application)ELISA
Quantity10 x 96 Tests
Unit SizeEach

Frequently asked questions (FAQs)

What substrate solution do you recommend using with IL-8 Human Matched Antibody Pair (Cat. No. MS204-3MST)?

We recommend the following formulation when making your own: 1:2 mixture of H2O2 and tetramethylbenzidine.
However we also offer a few alternatives for purchase:
- Pierce TMB Substrate Kit (Cat. No. 34021)
- Support Pack Standard (Cat. No. BMS412) and Support Pack Plus (Cat. No. BMS414). Both of these ELISA Support Packs include stabilized TMB substrate (does not require mixing), assay buffer, wash buffer, stop solution and PBS. Support Pack Plus includes plates.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.