Sodium hydroxide solution, 10N (30%), GMP
BioSpectra

Sodium hydroxide solution, 10N (30%), GMP

Low chloride, low iron, BET-tested, GMP manufactured bio pharma grade sodium hydroxide with WFI, this item is suitable for useRead more
Have Questions?
Change viewbuttonViewtableView
Quantity:
3.8 L
10 L
19 L
6 x 1 L
200 L
940 L
Catalog number BNH4150-10L
Price (INR)
-
Quantity:
10 L
Low chloride, low iron, BET-tested, GMP manufactured bio pharma grade sodium hydroxide with WFI, this item is suitable for use as a process chemical. It is manufactured in accordance with the IPEC-PQG Joint Good Manufacturing Practice Guide. This grade of sodium hydroxide solution is not suitable to be used as an active pharmaceutical ingredient, drug product, or household item.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point105–142°C
CAS1310-73-2
Chemical Name or MaterialSodium Hydroxide
ColorColorless
Molecular FormulaNaOH
PackagingCarboy
SynonymCaustic Soda, Lye, Sodium Hydrate
Molecular Weight (g/mol)40
Quantity10 L
pH14
Unit Size10 L