TBX Medium
Thermo Scientific™

TBX Medium

Detect E. coli in foods using this medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormat
BO0194M99-101 g125 mL sirop bottle
BO0194Z450 to 454 g500 mL DIN Bottle
Catalog number BO0194M
Price (INR)
-
Fill Weight:
99-101 g
Format:
125 mL sirop bottle
Tryptone Bile Glucuronide (TBX) Medium is based on Tryptone Bile Agar, which was originally formulated to improve on earlier methods used to detect E. coli in foods in terms of speed, reliability, better recovery from frozen samples and the detection of poor lactose fermenters
  • Tryptone Bile Glucuronide (TBX) Medium is based on Tryptone Bile Agar, which was originally formulated to improve on earlier methods used to detect E. coli in foods in terms of speed, reliability, better recovery from frozen samples and the detection of poor lactose fermenters
Specifications
ColorStraw
Fill Weight99-101 g
Format125 mL sirop bottle
Quantity10 x 100 mL
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
pH10 sirop bottles with screw cap closures in a box.
Unit SizeEach
Contents & Storage
2 to 10°C