Buffered Peptone Water (Bottled)
Thermo Scientific™

Buffered Peptone Water (Bottled)

Pre-enrich food samples prior to the isolation of Salmonella species using these ready-prepared media bottles.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightPackaging Type
BO0201S225.0g - 229.0g10 bottles with screw cap closures in a box
BO0201Z90g + 1.5g10 bottles with screw cap closures in a box
BO0201M99 to 101 g10 Sirop Bottles with screw cap closures in a box
BO0688S225 to 229 g10 wide-necked pots with screw cap closure in a box
EB0201E10.0 to 10.3 g100 universal bottles with screw cap closure in a box
Catalog number BO0201S
Price (INR)
-
Fill Weight:
225.0g - 229.0g
Packaging Type:
10 bottles with screw cap closures in a box

Pre-enrich food samples prior to the isolation of Salmonella species with Thermo Scientific™ Oxoid™ Buffered Peptone Water ready-prepared bottled medium. Buffered Peptone Water is a non-selective pre-enrichment medium.

Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.

  • Convenient: the pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.
  • Aids recovery of damaged Salmonella cells.

The test sample is added to Buffered Peptone Water at a ratio of 1:10 and incubated at 37° ± 1°C for 18 hours + 2 hours.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

Specifications
ColorStraw, Clear
DescriptionBuffered Peptone Water(Bottled)
Fill Weight225.0g - 229.0g
FormatSirop Bottle
Packaging Type10 bottles with screw cap closures in a box
Quantity10/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2 to 25°C