Tryptone Soya Agar with 1% Tween
Thermo Scientific™

Tryptone Soya Agar with 1% Tween

Recover and enumerate microorganisms from surfaces, materials, or products, especially after cleaning or disinfection procedures
Have Questions?
Catalog NumberFill Weight
BO0288V500 to 504 g
Catalog number BO0288V
Price (INR)
-
Fill Weight:
500 to 504 g
Tryptone Soya Agar with 1% Tween is for recovery and enumeration of microorganisms from surfaces, materials, or products, especially after cleaning or disinfection procedures.
Specifications
ColorStraw
Fill Weight500 to 504 g
Format500 mL Sirop Bottle
Packaging Type10 Sirop Bottles with screw cap closures in a box
Quantity10/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 25°C