Oxoid™ Buffered Sodium Chloride Peptone Solution (Bottled)
Thermo Scientific™

Oxoid™ Buffered Sodium Chloride Peptone Solution (Bottled)

Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP/IP with this isotonic medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill Weight
BO0322Z300 to 304 g
BO0322J90 to 91.5 g
BO0322X450 to 454 g
BO0322D9.0g + 0.3g
BO0322M100-101.5g
Catalog number BO0322Z
Price (INR)
-
Fill Weight:
300 to 304 g

Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP/IP with Thermo Scientific™ Buffered Sodium Chloride Peptone Solution. The combination of phosphate buffer, sodium chloride and peptone increases the viability of sensitive microorganisms by providing an isotonic environment. This is of particular importance when trying to recover cells that may be stressed or sensitive to osmotic pressure.

Use Buffered Sodium Chloride Peptone Broth for the dilution of samples in the analysis of non-sterile products for microbial contaminants.

  • Conforms to the harmonised Microbial Limit Testing chapters from the EP/USP/JP1,2,3
  • Maintains cell viability during dilution process.
  • Ready-to-use convenience of prepared media

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.

General References:

  1. US Pharmacopoeia 30 (2008)
  2. European Pharmacopoeia 6.1. 2.6.12. Microbiological Examination of Non-Sterile Products: Harmonised Method: Microbial Enumeration tests. 2.6.13. Microbiological Examination of Non-Sterile Products: Test for Specified microorganisms. B. Harmonised Method. (2008).
  3. Japanese Pharmacopoeia. 15 th Edition. (2006)
Specifications
ColorColorless
Fill Weight300 to 304 g
Format300 mL Sirop Bottle
Packaging Type10 Sirop Bottles with screw cap closure in a box
Quantity10/Pk.
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2° to 25°C