Tryptone Soya Agar
Tryptone Soya Agar
Thermo Scientific™

Tryptone Soya Agar

A non-selective medium recommended for use in qualitative procedures for the isolation and cultivation of a wide variety of organisms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormat
BO0330T248 to 252 g300 mL Sirop Bottle
BO0330M99-101g 125mL sirop bottle
BO0330V500 to 504 g500 mL Sirop Bottle
BO0405F15 to 15.3 gUniversal Bottle
Catalog number BO0330T
Price (INR)
-
Request A Quote
Fill Weight:
248 to 252 g
Format:
300 mL Sirop Bottle
Tryptone Soya Agar slopes may be used to cultivate, store, and ship bacterial cultures.
Specifications
ColorStraw
Fill Weight248 to 252 g
Format300 mL Sirop Bottle
Packaging Type10 Sirop Bottles with Screw Cap Closures/Box
Quantity10/Pk.
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 25°C