Membrane Lauryl Sulphate Broth
Membrane Lauryl Sulphate Broth
Thermo Scientific™

Membrane Lauryl Sulphate Broth

Selectively detect and enumerate coliform bacteria in water or food samples via membrane filtration.
Have Questions?
Catalog NumberFill Weight
BO0394E10.0 to 10.3 g
Catalog number BO0394E
Price (INR)
-
Request A Quote
Fill Weight:
10.0 to 10.3 g
Selectively detect and enumerate coliform bacteria in water or food samples via membrane filtration using Membrane Lauryl Sulphate Broth.
Specifications
ColorRed
Fill Weight10.0 to 10.3 g
FormatUniversal bottle
Packaging Type24 Universal Bottles with screw cap closures in a box
Quantity24/Pk.
Shelf Life35 weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 10°C