Oxoid™ Sabouraud Dextrose Agar
Thermo Scientific™

Oxoid™ Sabouraud Dextrose Agar

Sabouraud Dextrose Agar contains mycological peptone and glucose to support growth of fungi.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormat
BO0408V500-504 g 500 mL sirop bottle
BO0408T248 to 252 g300 mL Sirop Bottle
Catalog number BO0408V
Price (INR)
-
Fill Weight:
500-504 g
Format:
500 mL sirop bottle
The media are slightly acidic, which helps to inhibit bacteria while being favorable to fungi.
  • Sabouraud Dextrose Agar 10 x 500 mL: contain mycological peptone and glucose to support growth of fungi. The media are slightly acidic which helps to inhibit bacteria whilst being favourable to fungi
Specifications
ColorStraw
Fill Weight500-504 g
Format500 mL sirop bottle
Packaging Type10 sirop bottles with screw cap closures in a box.
Quantity10 x 500 mL
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2 to 25°C