This product is no longer available to order. If you would like to discuss your options, or ask for a technical representative to call you, please contact Technical Support.
Oxoid™ Enterobacteria Enrichment Broth-Mossel
Actual product may vary
Thermo Scientific™

Oxoid™ Enterobacteria Enrichment Broth-Mossel

A selective enrichment broth for the detection of bile tolerent Gram-negative bacteria in microbiological examination of pharmaceutical products complying with EP/USP/JP/BP specification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormatQuantity
BO1156J100 – 101.5 g100 mL sirop bottle10 x 90 mL
BO1156E10.0 to 10.3 gUniversal Bottle24/Pk.
BO1156M100-101.5 g 125 mL sirop bottle 10 x 100 mL
Catalog number BO1156J
Price (INR)
-
Fill Weight:
100 – 101.5 g
Format:
100 mL sirop bottle
Quantity:
10 x 90 mL
Bottled media for Enterobacteria enrichment media complying with EP/USP/JP/BP specification.
Specifications
ATCC®ARTG
Certifications/ComplianceEP/USP/JP/BP
Country of OriginUK
Fill Weight100 – 101.5 g
For Use With (Application)Enrichment Broth for Enterobacteria
Format100 mL sirop bottle
Media TypePrepared Media
Packaging Type10 sirop bottles bottles with screw cap closures in a box
Quantity10 x 90 mL
Shelf Life6 - 12 months
Product TypeMicrobiology Media
SpeciesEnterobacteria
Unit SizeEach
Contents & Storage
2°C to 8°C