Oxoid™ Enterobacteria Enrichment Broth-Mossel
Thermo Scientific™

Oxoid™ Enterobacteria Enrichment Broth-Mossel

A selective enrichment broth for the detection of bile tolerent Gram-negative bacteria in microbiological examination of pharmaceutical products complying with EP/USP/JP/BP specification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormatQuantity
BO1156M100-101.5 g 125 mL sirop bottle 10 x 100 mL
BO1156J100 – 101.5 g100 mL sirop bottle10 x 90 mL
BO1156E10.0 to 10.3 gUniversal Bottle24/Pk.
Catalog number BO1156M
Price (INR)
-
Fill Weight:
100-101.5 g
Format:
125 mL sirop bottle
Quantity:
10 x 100 mL
Bottled media for Enterobacteria enrichment media complying with EP/USP/JP/BP specification.
Specifications
ATCC®ARTG
Certifications/ComplianceEP/USP/JP/BP
ColorGreen
Fill Weight100-101.5 g
Format125 mL sirop bottle
Packaging Type10 sirop bottles with screw cap closures in a box.
Quantity10 x 100 mL
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeBroth
Unit SizeEach
Contents & Storage
2 to 25°C