Contrast™ MRSA Broth, 3 ± 0,3mL
Product Image
Thermo Scientific™
Contrast™ MRSA Broth, 3 ± 0,3mL
Improve laboratory productivity by adding multiple clinical samples from one patient to one tube and achieve results within 18 to 24 hours for methicillin-resistant Staphylococcus aureus (MRSA).
Have Questions?
Catalog number BO1226B
Price (INR)
-
Request A Quote
Improve laboratory productivity by adding multiple samples from one patient to one tube and achieve results within 18 to 24 hours with Thermo Scientific™ Contrast™ MRSA Broth. Rapidly eliminate negative results and confirm positive results using Thermo Scientific™ Brilliance™ MRSA 2 Agar.
Simplicity and speed are important factors when screening for MRSA, as reliable results are necessary for an effective infection control process.
Contrast MRSA Broth contains the same antibiotic marker used in Brilliance MRSA Agar to select for MRSA strains. Other antibiotics and inhibitory agents are present to inhibit non-MRSA and Gram-negative organisms. The broth contains an indicator dye, phenol red, and two sugars (mannitol and trehalose) which are fermented by MRSA strains. This causes a decrease in the pH of the broth, resulting in a color change from red to orange/yellow, indicating a presumptive positive result. The improved broth contains a low level of nalidixic acid, which has activity against both Gram-positive and Gram-negative bacteria. The concentration is optimized for maximum activity against non-target organisms without compromising growth of methicillin-resistant S. aureus, including ciprofloxicin-sensitive strains. Contrast MRSA Broth can be inoculated from screening swabs taken from hospital patients or staff, according to local guidelines. Multiple swabs (maximum four) from the same patient can be pooled into a single broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
Specifications
ColorRed, clear
DescriptionContrast MRSA Broth
Fill Volume3 mL
Fill Weight3 ± 0,3mL
FormatUniversal
Packaging Type24 universal in a carton box
Quantity24/Pk.
Shelf Life6 Weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach