Buffered Peptone Water
Thermo Scientific™

Buffered Peptone Water

Pre-enrich food samples prior to the isolation of Salmonella species with this non-selective medium while adhering to ISO 11133:2014.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormat
BO1277S225 to 227 g500 mL HDPE Sirop Bottle
BO1067S223-227 g225 mL sirop bottle
EB1067Z950 to 955 g1 L Sirop Bottle
Catalog number BO1277S
Price (INR)
-
Fill Weight:
225 to 227 g
Format:
500 mL HDPE Sirop Bottle
Pre-enrich food samples prior to the isolation of Salmonella species with Buffered Peptone Water.
Specifications
ColorStraw
Fill Weight225 to 227 g
Format500 mL HDPE Sirop Bottle
Packaging Type10 HDPE Sirop Bottles with Screw Cap Closures/Box
Quantity10/Pk.
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C