Modified Tryptone Soya Broth
Thermo Scientific™

Modified Tryptone Soya Broth

Selectively enrich and isolate Escherichia coli O157 from food with this ready to use medium.
Have Questions?
Catalog NumberFill Weight
BO1315S225 to 229 g
Catalog number BO1315S
Price (INR)
-
Fill Weight:
225 to 229 g
Selectively enrich and isolate Escherichia coli O157 from food with Modified Tryptone Soya Broth.
Specifications
ColorStraw
Fill Weight225 to 229 g
Format250 mL Sirop Bottle
Packaging Type10 Sirop Bottles with screw cap closures in a box
Quantity10/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C