BPC ReadyMate™ to Steam Through Tubing Set
BPC ReadyMate™ to Steam Through Tubing Set
BPC ReadyMate™ to Steam Through Tubing Set
BPC ReadyMate™ to Steam Through Tubing Set
Thermo Scientific™
BPC ReadyMate™ to Steam Through Tubing Set
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Multiple connections give greater flexibility.
Have Questions?
Catalog number BP0070A
Price (INR)
-
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Create a sterile union to allow additional aseptic or steam connections using the Thermo Scientific™ BPC ReadyMate™ to Steam Through Tubing Set.
  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labor costs
Multiple plug-and-play connection options, such as BPC ReadyMate to Steam Through, are available for greater flexibility.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.
Specifications
ColorMulti
DescriptionBPC ReadyMate to Steam Through Tubing Set
FormatBioprocess Contaners (prepared media)
IncludesTubing and connectors
Length (Metric)1 m
Quantity1/Pk.
Product TypeMicrobiology Media
Unit SizeEach