BPC ReadyMate™ to 2X ReadyMate Y Piece Tubing Set
Thermo Scientific™

BPC ReadyMate™ to 2X ReadyMate Y Piece Tubing Set

Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Multiple connections give greater flexibility.
Have Questions?
Catalog NumberFormat
BP0080AY Piece Tubing Set
Catalog number BP0080A
Price (INR)
-
Format:
Y Piece Tubing Set
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Create a sterile union to allow additional aseptic or steam connections using the Thermo Scientific™ BPC ReadyMate™ to 2X ReadyMate Y Piece Tubing Set.
  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labor costs

Multiple plug-and-play connection options, such as BPC ReadyMate to 2X ReadyMate, are available for greater flexibility.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.

Specifications
DescriptionBPC ReadyMate to 2X ReadyMate Y Piece Tubing Set
FormatY Piece Tubing Set
IncludesTubing and connectors
Length (Metric)1 m
Quantity1/Pk.
Unit SizeEach