Teledyne CETAC Autosamplers
Teledyne CETAC Autosamplers
Teledyne CETAC Autosamplers
Teledyne CETAC Autosamplers
Teledyne CETAC Autosamplers
Teledyne CETAC Autosamplers
Thermo Scientific™
Teledyne CETAC Autosamplers
Increase sample throughput with advanced Teledyne CETAC Autosamplers, compatible with Thermo Scientific™ high-performance trace elemental analyzers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Model
BRE0003260iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MSCETAC ASX-560 Autosampler
BRE0007611iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MSCETAC ASX-280 Autosampler
BRE0013184iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MSCETAC XLR-860 Autosampler
BRE0018963iCAP Series ICP-OESCETAC ASX7400 Stir Autosampler
Catalog number BRE0003260
Price (INR)
-
Request A Quote
For Use With (Equipment):
iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MS
Model:
CETAC ASX-560 Autosampler
Increase sample throughput in your laboratory with Teledyne CETAC Autosamplers. This selection of advanced autosamplers, compatible with Thermo Scientific™ high performance trace elemental analyzers, enables operators to take on other tasks, while intelligent, automated sample introduction accelerates workflows and eliminates manual errors.
Teledyne CETAC ASX-280 Autosampler
Accommodates up to 180 sample capacity for low to moderate sample throughput, compatible with:
  • Thermo Scientific™ iCE™ 3000 Series AAS systems
  • Thermo Scientific™ iCAP™ 7000 Series ICP-OES systems
  • Thermo Scientific™ iCAP™ PRO Series ICP-OES systems
  • Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS systems
  • Thermo Scientific™ Element™ Series HR-ICP-MS systems
Teledyne CETAC ASX-560 Autosampler
Accommodates up to 360 samples for moderate to high sample throughput, compatible with:
  • iCE 3000 Series AAS systems
  • iCAP 7000 Series ICP-OES systems
  • iCAP PRO Series ICP-OES systems
  • iCAP Q/Qnova Series ICP-MS systems
  • Element Series HR-ICP-MS systems
Teledyne CETAC XLR-860 Autosampler
Enables the unattended analysis of up to 720 samples, for laboratories looking to push the boundaries of unattended analysis, compatible with:
  • iCAP 7000 Series ICP-OES systems
  • iCAP PRO Series ICP-OES systems
  • iCAP Q/Qnova Series ICP-MS systems
Teledyne CETAC ASX-7400 Autosampler
Incorporates automated liquid stirring to ensure sample homogeneity, for samples containing particulates in suspension such as wear metals applications, compatible with:
  • iCAP 7000 Series ICP-OES systems
  • iCAP PRO Series ICP-OES systems
Specifications
ModelCETAC ASX-560 Autosampler
For Use With (Equipment)iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MS
TypeAutosampler
Unit SizeEach