2-Channel Peristaltic Pump Tubing
Thermo Scientific™

2-Channel Peristaltic Pump Tubing

Keep your Teledyne CETAC™ Autosampler system in optimum condition using dedicated consumables, including Teledyne CETAC™ Peristaltic Pump Tubing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeColorMaterial
BRE0004340Peristaltic Pump TubingBlackViton
BRE0004331Peristaltic Pump TubingTanPharMed
BRE0004334Peristaltic Pump TubingClearTygon
BRE0004337Peristaltic Pump TubingYellowTygon Fuels
Catalog number BRE0004340
Price (INR)
-
Type:
Peristaltic Pump Tubing
Color:
Black
Material:
Viton
Keep your Teledyne CETAC™ Autosampler system in optimum condition using dedicated consumables, including Teledyne CETAC™ 2-channel peristaltic pump tubing.

Ensure optimum flow of your rinse solution with replacement 2-channel peristaltic pump tubing. For use with pumps that use 2-channel tubing cartridges.

Material compatibilities with applications:

  • PharMed™—pharmaceutical, biological, food and beverage
  • Viton™—acids, solvents, oils, high-temperature applications
  • Tygon™—laboratory research, food and beverage, biological, pharmaceutical, medical device, brewery, consumer products, environmental, raw milk
  • Tygon™ Fuel and Lubricant—most fuels and industrial lubricants
Specifications
ColorBlack
Diameter (Metric) Inner2 x 2 mm
MaterialViton
For Use With (Equipment)Teledyne CETAC Autosamplers
TypePeristaltic Pump Tubing
Unit SizeEach