GCI Series Interface
Actual product may vary
GCI Series Interface
GCI Series Interface
Thermo Scientific™
GCI Series Interface
Link your GC and ICP-MS systems seamlessly using the Thermo Scientific™ GCI Series Interface for ultimate ease-of-use in advanced speciation analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
BRE0008121GCI 200-Interface ELEMENT 2/XR
BRE0008120GCI 100-Interface iCAP Q/Qnova
BRE0008122GCI 300-Interface NEPTUNE Plus
Catalog number BRE0008121
Price (INR)
-
Type:
GCI 200-Interface ELEMENT 2/XR
Use the Thermo Scientific™ GCI Series Interface to seamlessly integrate your gas chromatography (GC) and inductively coupled plasma mass spectrometry (ICP-MS) systems as an easy-to-use solution for advanced speciation studies. When GC and ICP-MS systems are seamlessly connected, even applications having the most challenging detection limit are no problem. The GCI Series Interface provides easy, reliable measurement of volatile species, down to sub-ppt concentrations.
Easy and simple
  • Plug and play device
  • Easily couples the Thermo Scientific™ TRACE™ 1310 GC oven with any of our Thermo Scientific™ ICP-MS instruments
  • Flexible design
  • Installs in less than 10 minutes thanks to a simple, gas-tight, push-fit connectivity with the ICP torch
Seamless integration
The GCI Series Interface provides:
  • An integrated parameter of your GC program, just like injector and autosampler
  • Higher sensitivity
  • Higher data confidence
  • Reliability
  • Homogeneous isothermal temperature profile—no cold or hot spots that could cause column damage or loss of the target compounds
  • Chromatographic reproducibility with baseline separated and quantifiable peaks in every run
  • GCI interface interlock and ultra-fast cooling/heating of the transferline to ensure safe and reliable operation of the system in any laboratory
Flexibility
  • Tailor your system to the needs of your application. With a range of TRACE 1300 and 1600 Series GC options, application specific columns and advanced sample handling functions, you can be sure to meet your application requirements.
  • A range of GC autosamplers gives you flexibility in performing liquid, solid and gas sampling with precision and reliability.
Specifications
For Use With (Equipment)ICP-MS Systems
TypeGCI 200-Interface ELEMENT 2/XR
Unit SizeEach