Teledyne CETAC SDX High Performance Liquid Dilution System
Teledyne CETAC SDX High Performance Liquid Dilution System
Thermo Scientific™
Teledyne CETAC SDX High Performance Liquid Dilution System
Reduce sample preparation time and errors with the Teledyne CETAC SDXHPLD automated dilution system, based on the robust, industry-standard Teledyne CETAC ASX-560 autosampler
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
BRE0008829iCAP PRO, Q/Qnova
BRE0008830Teledyne CETAC, iCAP Q/Qnova
Catalog number BRE0008829
Price (INR)
-
Request A Quote
For Use With (Equipment):
iCAP PRO, Q/Qnova
Reduce sample preparation time and errors with the simple, straightforward Teledyne CETAC SDXHPLD automated dilution system, based on the robust, industry-standard Teledyne CETAC ASX-560 Autosampler. The CETAC SDXHPLD uses volume-to-volume dilution with a highly accurate syringe pump and unique vortex mixing.
T​he CETAC SDXHPLD automated dilution system offers:
Prescriptive Dilution (2x to 5000x)
  • Saves laboratory staff time and effort
  • Provides accurate and precise unknown sample dilution
  • Offers calibration standard dilution
Responsive and Intelligent Dilution
  • Provides automated re-analysis and sample introduction for both samples exceeding calibration range and samples with suppressed internal standard response
Specifications
DescriptionComplete SDXHPLD Autodilution System including ASX-560 Autosampler, Vortex Mixer and Syringe Pump Module
For Use With (Equipment)iCAP PRO, Q/Qnova
TypeAutodilution System
Unit SizeEach