Teledyne CETAC SDX High Performance Liquid Dilution System
Thermo Scientific™
Teledyne CETAC SDX High Performance Liquid Dilution System
Reduce sample preparation time and errors with the Teledyne CETAC SDXHPLD automated dilution system, based on the robust, industry-standard Teledyne CETAC ASX-560 autosampler
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
BRE0008830
Teledyne CETAC, iCAP Q/Qnova
BRE0008829
iCAP PRO, Q/Qnova
2 Options
Catalog number BRE0008830
Price (INR)
-
Request A Quote
For Use With (Equipment):
Teledyne CETAC, iCAP Q/Qnova
Reduce sample preparation time and errors with the simple, straightforward Teledyne CETAC SDXHPLD automated dilution system, based on the robust, industry-standard Teledyne CETAC ASX-560 Autosampler. The CETAC SDXHPLD uses volume-to-volume dilution with a highly accurate syringe pump and unique vortex mixing.
The CETAC SDXHPLD automated dilution system offers:
Prescriptive Dilution (2x to 5000x)
Saves laboratory staff time and effort
Provides accurate and precise unknown sample dilution
Offers calibration standard dilution
Responsive and Intelligent Dilution
Provides automated re-analysis and sample introduction for both samples exceeding calibration range and samples with suppressed internal standard response
Specifications
DescriptionSDXHPLD Upgrade Kit for existing ASX-560 Autosampler, including Vortex Mixer and Syringe Pump Module only
For Use With (Equipment)Teledyne CETAC, iCAP Q/Qnova