Teledyne CETAC Autosampler Sample Trays
Teledyne CETAC Autosampler Sample Trays
Teledyne CETAC Autosampler Sample Trays
Teledyne CETAC Autosampler Sample Trays
Thermo Scientific™
Teledyne CETAC Autosampler Sample Trays
Make lab work easier and more productive with high-quality, dependable Teledyne CETAC autosampler accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeIncludes
BRE0022739CETAC ASX-7400 AutosamplerAutosampler Sample Tray
BRE0004343CETAC ASX-560 AutosamplerAutosampler Sample Tray
BRE0007612CETAC ASX-280 AutosamplerAutosampler Sample Tray
Catalog number BRE0022739
Price (INR)
-
For Use With (Equipment):
CETAC ASX-7400 Autosampler
Type:
Autosampler Sample Tray
Make lab work easier and more productive with high-quality, dependable Teledyne CETAC autosampler accessories. Teledyne CETAC Autosampler Sample Trays feature ribs on the bottom to hold the sample vial securely in place.
Match your Teledyne CETAC Autosampler Sample Tray to Your Laboratory Needs
  • Sample tray for Teledyne CETAC ASX-280 Autosampler has 10 rear standard positions and accommodates up to two sample vial racks
  • Sample tray for Teledyne CETAC ASX-560 Autosampler has 10 rear standard positions and accommodates up to four sample vial racks
  • Sample tray for Teledyne CETAC ASX-7400 Stirring Autosampler accommodates up to two standard racks with 5 positions each and up to four sample vial racks
Specifications
For Use With (Equipment)CETAC ASX-7400 Autosampler
TypeAutosampler Sample Tray
Unit SizeEach