Single-Channel Peristaltic Pump Tubing
Actual product may vary
Single-Channel Peristaltic Pump Tubing
Single-Channel Peristaltic Pump Tubing
Single-Channel Peristaltic Pump Tubing
Single-Channel Peristaltic Pump Tubing
Thermo Scientific™

Single-Channel Peristaltic Pump Tubing

Keep your Teledyne CETAC™ Autosampler system in optimum condition using dedicated consumables, including Teledyne CETAC™ single-channel peristaltic pump tubing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDiameter (Metric) InnerColorMaterial
BRE0027233Peristaltic Pump Tubing2 mmBlackViton
BRE0027227Peristaltic Pump Tubing2 mmTanPharMed
BRE0027229Peristaltic Pump Tubing2 mmClearTygon
Catalog number BRE0027233
Price (INR)
-
Type:
Peristaltic Pump Tubing
Diameter (Metric) Inner:
2 mm
Color:
Black
Material:
Viton
Keep your Teledyne CETAC™ Autosampler system in optimum condition using dedicated consumables, including Teledyne CETAC™ single-channel peristaltic pump tubing.

Ensure optimum flow of your rinse solution with replacement peristaltic single-channel pump tubing. For use with pumps that use single-channel tubing cartridges

Material compatibilities with applications:

  • PharMed™: pharmaceutical, biological, food and beverage
  • Viton™: acids, solvents, oils, high-temperature applications
  • Tygon™: laboratory research, food and beverage, biological, pharmaceutical, medical device, brewery, consumer products, environmental, raw milk
  • Tygon™ Fuel and Lubricant: most fuels and industrial lubricants
Specifications
ColorBlack
Diameter (Metric) Inner2 mm
MaterialViton
For Use With (Equipment)Teledyne CETAC Autosamplers
TypePeristaltic Pump Tubing
Unit SizeEach