Sample Loops for ESI FAST and prepFAST Autosamplers
Thermo Scientific™
Sample Loops for ESI FAST and prepFAST Autosamplers
Optimize autosampler throughput with precision sample loops for ESI FAST and prepFAST systems, delivering consistent volumes and reliable, leak‑tight performance.
Have Questions?
Catalog NumberType
BRE0029725Aqueous Sample Loop
Catalog number BRE0029725
Price (INR)
-
Type:
Aqueous Sample Loop
Boost consistency and throughput with these sample loops for ESI FAST and prepFAST Autosamplers, engineered to deliver accurate, repeatable volumes for stable sample transfer. These direct‑fit loops support rapid switching, preserve leak‑tight connections, and simplify routine replacement to keep workflows efficient and instruments ready.
Increase throughput
  • Enable rapid, reliable loop loading to keep FAST and prepFAST systems delivering samples efficiently.
Ensure volume accuracy
  • Provide repeatable loop volumes
  • Support consistent sample transfer.
Install easily
  • Use direct‑fit designs
  • Simplify routine replacement.
Specifications
Diameter (Metric)3.2 mm
Diameter (Metric) Inner1.6 mm
Volume (Metric)1 mL
For Use With (Equipment)ESI FAST, ESI prepFAST 2
TypeAqueous Sample Loop
Unit SizeEach