iSC-65 Autosampler Probes
Thermo Scientific™

iSC-65 Autosampler Probes

Maximize the versatility and efficiency of your laboratory with Thermo Scientific™ iSC-65 Autosampler Probes, designed for diverse applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDiameter (Metric) InnerFor Use With (Application)
BRE0030312Blue Band0.50 mmICP-MS
BRE0030311Black Band0.25 mmLow Flow Rates
BRE0030313Red Band0.75 mmICP-OES
BRE0030314Green Band1.00 mmHigh Flow Rates
Catalog number BRE0030312
Price (INR)
-
Color:
Blue Band
Diameter (Metric) Inner:
0.50 mm
For Use With (Application):
ICP-MS
Easy to install and compatible with the iSC-65 Autosampler system, customized iSC-65 Autosampler sample probes enhance productivity and consistency in your workflows, making them essential for achieving optimal results for trace elemental analysis.

Customized autosampler accessories ensure more efficient, reliable analysis for your autosampler and trace elemental analyzer.

Choose an iSC-65 Autosampler sample probe from the following options:

  • 0.25 mm ID—used for low-flow applications
  • 0.50 mm ID—typically used with ICP-MS systems
  • 0.75 mm ID—typically used with ICP-OES systems
  • 1.00 mm ID—used for high-flow applications
Specifications
ColorBlue Band
Diameter (Metric) Inner0.50 mm
For Use With (Application)ICP-MS
MaterialPTFE-sheathed Carbon Fiber
For Use With (Equipment)iSC-65 Autosampler
TypeAutosampler Sample Probe
Unit SizeEach