iSC-65 Rinse and Drain Tubing Kit
Thermo Scientific™
iSC-65 Rinse and Drain Tubing Kit
Enhance performance and safety in your laboratory and boost efficiency with the Thermo Scientific™ iSC-65 Rinse and Drain Tubing Kit. Ensure consistent results and reduce contamination risks.
Have Questions?
Catalog NumberType
BRE0032455Rinse and Drain Tubing Kit
Catalog number BRE0032455
Price (INR)
-
Type:
Rinse and Drain Tubing Kit
Designed for seamless integration with the iSC-65 Autosampler system, this kit ensures efficient rinsing and drainage, reducing the risk of contamination and maintaining sample integrity. Made from high-quality, durable materials, it offers reliable and long-lasting use.
Enhanced Laboratory Efficiency
  • Optimizes rinsing and drainage processes
  • Reduces downtime with quick and easy installation
  • Supports consistent and accurate sample handling
Specifications
MaterialTygon
For Use With (Equipment)iSC-65 Autosampler
TypeRinse and Drain Tubing Kit
Unit SizeEach