Liquid Sample Introduction Kits for iCAP™ PRO Series ICP-OES Systems
Liquid Sample Introduction Kits for iCAP™ PRO Series ICP-OES Systems
Thermo Scientific™
Liquid Sample Introduction Kits for iCAP™ PRO Series ICP-OES Systems
Deliver increased flexibility and precise sample introduction with preconfigured Thermo Scientific™ Liquid Sample Introduction Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
BRE0063034Duo Electrolyte Kit
BRE0019681Duo Aqueous Kit
BRE0019682Duo High Solids Kit
BRE0019683Duo Organics Kit
BRE0019684Radial Volatile Organics Kit
BRE0019685Radial HF Kit
BRE0019956Radial Aqueous Kit
BRE0019957Radial High Solids Kit
BRE0019958Radial Organics Kit
BRE0019959Duo Volatile Organics Kit
BRE0019960Duo HF Kit
BRE0063033Duo Cathode Kit
Catalog number BRE0063034
Price (INR)
-
Type:
Duo Electrolyte Kit
Increase your flexibiity to analyze a wide variety of sample matrices using this range of preconfigured, plug-and-play Thermo Scientific™ Liquid Sample Introduction Kits for use with Thermo Scientific™ iCAP™ PRO Series ICP-OES systems. Optional, dedicated kits are available for samples with high solid content or those containing organic, volatile organic or hydrofluoric acid (HF). These kits offer precise, accurate sample delivery every time.
Increase flexibility with application-specific sample introduction kits
We offer a variety of sample introduction kits for ICP-OES applications, including:
  • Aqueous - ideal for liquid sample solutions containing low acid content and low dissolved salts, this kit is supplied as standard
  • High Solids - necessary to prevent blockages when a sample includes solid particles in suspension
  • Organics - recommended for samples containing relatively non-volatile, organic solvents, such as xylene, toluene, white spirit, and kerosene
  • Volatile Organics* - reduces the plasma solvent load for optimal analysis of highly volatile solvents
  • HF - effectively handles hydrofluoric acid (HF) sample solutions
Boost performance with precision sample introduction kits
Just like our analyzers, our sample introduction kits are designed to maximize convenience, ease of use, and precision.
  • All sample introduction kits are plug-and-play and self-aligning for convenience in routine maintenance
  • Semi-demountable torch module simply clips into place and automatically interlocks with the system for gas-tight connections
  • Center tube is removable with the torch in situ for dramatically reduced down-time during maintenance and cleaning
Ordering RequirementsPeristaltic pump tubing sold separately.
* A Peltier-cooled spray chamber like the IsoMist XR needs to be purchased in addition to the Volatile Organics sample introduction kit.
Specifications
For Use With (Equipment)iCAP PRO XP/XPS ICP-OES Duo
TypeDuo Electrolyte Kit
Unit SizeEach