Cyclonic Spray Chambers
Thermo Scientific™
Cyclonic Spray Chambers
Low-volume, baffled cyclonic spray chambers efficiently filter out larger aerosol droplets for improved plasma stability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
BRE0063530Spray Chamber Exit PortGlass
1317080Cyclonic Spray ChamberQuartz
1320260Cyclonic Spray ChamberPFA
BRE0061134Spray Chamber Exit PortPFA
BRE0061036iCAP MX Spray ChamberGlass
BRE0032947iCAP MX Spray ChamberPFA
Catalog number BRE0063530
Price (INR)
-
Type:
Spray Chamber Exit Port
Material:
Glass
Available in quartz or PFA to suit your analysis requirements, these Thermo Scientific™ Cyclonic Spray Chambers are for use with the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems. Low-volume, baffled cyclonic spray chambers efficiently filter out larger aerosol droplets for improved plasma stability.
Ensure optimal ICP-MS instrument performance
Spray chambers for ICP-MS instruments ensure efficient atomization and ionization of the sample for accurate analysis
  • High-purity quartz spray chambers are suited to standard ICP-MS applications, except for introduction HF and highly concentrated acids
  • PFA spray chambers are suitable for ultra-trace analysis, introduction of HF and highly concentrated acids
Specifications
For Use With (Equipment)iCAP MX Series ICP-MS Instruments
MaterialGlass
TypeSpray Chamber Exit Port
Unit SizeEach