Orbitrap™ Exploris™ GC S Mass Spectrometer
Orbitrap™ Exploris™ GC S Mass Spectrometer
Orbitrap™ Exploris™ GC S Mass Spectrometer
Orbitrap™ Exploris™ GC S Mass Spectrometer
Orbitrap™ Exploris™ GC S Mass Spectrometer
Orbitrap™ Exploris™ GC S Mass Spectrometer
Orbitrap™ Exploris™ GC S Mass Spectrometer
Thermo Scientific™
Orbitrap™ Exploris™ GC S Mass Spectrometer
Modernize GC-MS analysis with high-resolution accuracy for dioxins, pesticides, and POPs—delivering regulatory-ready results with flexible scope expansion and simplified workflows.
Have Questions?
Catalog number BRE725547
Price (INR)
-
Modernize your dioxin and POPs testing with the Orbitrap Exploris GC S, engineered to expand analytical scope while delivering confident, defensible results. Achieve unmatched sensitivity, selectivity, and robustness in a compact, easy-to-use system that maximizes throughput, reduces costs, and empowers labs to meet demanding targets with precision and reliability.
Deliver accurate quantitative results
  • Full-scan HRAM data supports quantitation, screening, and confirmation
  • Reliable low-level performance across food and environmental matrices
  • Reduced data review through selective HRAM detection.
Increase selectivity in complex samples
  • Multi-point identification using accurate mass, isotope patterns, retention indices, and confirmation ions
  • Improved discrimination from matrix interferences with 60,000 mass resolving power (FWHM m/z 200)
  • Regulatory-ready identification for dioxins, pesticides, and POPs.
Stay ahead of evolving demands
  • Expand analytical scope without system reconfiguration
  • Support method consolidation across compound classes
  • Protect long-term investment as regulations evolve.
Focus on results, not instrument complexity
  • Rapid deployment with simple system setup
  • Chromeleon™ CDS Dioxin HRAM eWorkflow guide compliant execution and reporting
  • Standardized workflows reduce reliance on specialist expertise.
Specifications
Mass AccuracyExternal calibration achieves <3 ppm RMS drift over 24 hours, Internal lock mass calibration achieves <1 ppm RMS drift over 24 hours
ModelOrbitrap Exploris GC S MS
Scan RatesUp to 40 Hz at resolution setting 7,500 at m/z 200
Mass Range30 to 3000 m/z
Unit SizeEach