Trehalose Dihydrate, LBLE, EP, JP, NF, GMP
Trehalose Dihydrate, LBLE, EP, JP, NF, GMP
BioSpectra

Trehalose Dihydrate, LBLE, EP, JP, NF, GMP

Low bioburden, low endotoxin, EP, JP, NF, GMP manufactured, excipient grade trehalose dihydrate. This item is suitable for use asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
BTRED-3252-2525 kg
BTRED-3252-1010 kg
Catalog number BTRED-3252-25
Price (INR)
-
Request A Quote
Quantity:
25 kg
Low bioburden, low endotoxin, EP, JP, NF, GMP manufactured, excipient grade trehalose dihydrate. This item is suitable for use as an excipient, and is manufactured in accordance with the ICH-Q7 Good Manufacturing Practice Guide. This grade of trehalose dihydrate is not suitable to be used as an active pharmaceutical ingredient, drug product, or household item.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorColorless
PackagingHDPE round pail with lid, LDPE liner
GradeEP, JP, NF
Molecular Weight (g/mol)378.33
Percent Purity98.0–101.0%
Physical FormLiquid
Quantity25 kg
pH4.5–6.5
Unit Size25 kg