Tromethamine (Tris base), USP, EP, JP, LBLE, GMP, Excipient Grade
BioSpectra

Tromethamine (Tris base), USP, EP, JP, LBLE, GMP, Excipient Grade

Low bioburden, low endotoxin, multicompendial, GMP manufactured, bio excipient grade tromethamine (Tris base). This product is suitable for use asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
BTRIS-3255-1010 kg
BTRIS-3255-2525 kg
BTRIS-3255-5050 kg
Catalog number BTRIS-3255-10
Price (INR)
-
Quantity:
10 kg
Low bioburden, low endotoxin, multicompendial, GMP manufactured, bio excipient grade tromethamine (Tris base). This product is suitable for use as an excipient, and is manufactured in accordance with the ICH-Q7 Good Manufacturing Practice Guide. This grade of tromethamine (Tris base), multicompendial is not suitable to be used as an active pharmaceutical ingredient, drug product, or household item.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point220°C
CAS77-86-1
Chemical Name or MaterialTromethamine (Tris base)
ColorWhite
Molecular FormulaC4H11NO3
PackagingHDPE round pail with lid, LDPE liner
SynonymTrometamol, THAM, Tris (Hydroxymethyl) Aminomethane, Trisaminol, Trismethylolaminomethane
Molecular Weight (g/mol)121.14
Percent Purity99.0–101.0%
Physical FormCrystalline Powder
Quantity10 kg
pH10–11.5
Unit Size10 kg