Bandmate™ Blot Tray Cover
Invitrogen™
Bandmate™ Blot Tray Cover
The Bandmate Blot Tray Cover is a replacement cover specially designed for use with the Bandmate Automated Western Blot ProcessorRead more
Have Questions?
Catalog NumberQuantity
BW00501 Tray Cover
Catalog number BW0050
Price (INR)
-
Quantity:
1 Tray Cover
The Bandmate Blot Tray Cover is a replacement cover specially designed for use with the Bandmate Automated Western Blot Processor mini and midi incubation trays. This tray cover is required to protect blots and reagents during western blot processing.
Learn more about the Bandmate Automated Western Blot Processor ›
Specifications
ColorClear
DescriptionBandmate Tray Cover
Dimensions (L x W x H)21 cm x 12.7 cm x 0.953 cm
Quantity1 Tray Cover
Shipping ConditionAmbient
Weight0.091 kg
Weight (English)3.2 oz
Weight (Metric)0.091 kg
For Use With (Application)Western Blot
For Use With (Equipment)Invitrogen Bandmate Automated Western Blot Processor
Product LineBandmate
TypeTray Cover
Unit SizeEach