Bandmate™ Reagent Tubes
Invitrogen™
Bandmate™ Reagent Tubes
Bandmate Reagent Tubes are specially designed for use with the Bandmate Automated Western Blot Processor. The tubes are required forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
BW020X5100 tubes
BW020X25500 tubes
Catalog number BW020X5
Price (INR)
11,220.00
Online offer
Ends: 31-Mar-2026
16,621.00
Save 5,401.00 (32%)
Each
Quantity:
100 tubes
Price (INR)
11,220.00
Online offer
Ends: 31-Mar-2026
16,621.00
Save 5,401.00 (32%)
Each
Bandmate Reagent Tubes are specially designed for use with the Bandmate Automated Western Blot Processor. The tubes are required for dispensing primary and secondary antibodies into the Bandmate mini and midi incubation trays during the automated western blot processing procedure. Two reagent tubes are required for each blot.
Learn more about the Bandmate Automated Western Blot Processor ›
Specifications
ColorClear
DescriptionBandmate Reagent Tubes, 100 Pack
Quantity100 tubes
Shipping ConditionAmbient
For Use With (Application)Western Blot
For Use With (Equipment)Invitrogen Bandmate Automated Western Blot Processor
Product LineBandmate
TypeReagent Tube
Unit SizeEach