Coverglass Removal Tool
Invitrogen™
Coverglass Removal Tool
The Coverglass Removal Tool is intended for use with a CultureWell™ Chambered Coverglass for Cell Culture (Cat. Nos. C37000, C37005).Read more
Have Questions?
Catalog NumberQuantity
C370101 each
Catalog number C37010
Price (INR)
3,822.00
Each
Quantity:
1 each
Price (INR)
3,822.00
Each
The Coverglass Removal Tool is intended for use with a CultureWell™ Chambered Coverglass for Cell Culture (Cat. Nos. C37000, C37005). This tool enables you to easily separate the chambered upper structure from the coverglass without the need for excessive force, minimizing the risk of coverglass breakage.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Microscope
Quantity1 each
Product LineCulturewell
TypeCoverglass Removal Tool
Unit SizeEach