Assembled Target™ 10-425 Caps and Septa
Assembled Target™ 10-425 Caps and Septa
Actual product may vary
Assembled Target™ 10-425 Caps and Septa
Assembled Target™ 10-425 Caps and Septa
Assembled Target™ 10-425 Caps and Septa
Thermo Scientific™

Assembled Target™ 10-425 Caps and Septa

Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.
Have Questions?
Catalog NumberColorSeptum
C4010-5ANaturalIntegral Molded Polypropylene
Catalog number C4010-5A
Price (INR)
-
Request A Quote
Color:
Natural
Septum:
Integral Molded Polypropylene

Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.

  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Storage cap has a solid top and pre-inserted PTFE disk liner
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure Size10 mm
DescriptionNatural; Septa: Integral Molded Polypropylene
For Use With (Equipment)10-425 Screw Thread Vials
SeptumIntegral Molded Polypropylene
ColorNatural
MaterialPolypropylene
Product LineNational Assembled Target
TypeScrew Cap
Unit SizePack of 100