Lindberg/Blue M™ Box and Tube Furnace Controllers
Lindberg/Blue M™ Box and Tube Furnace Controllers
Actual product may vary
Lindberg/Blue M™ Box and Tube Furnace Controllers
Lindberg/Blue M™ Box and Tube Furnace Controllers
Thermo Scientific™
Lindberg/Blue M™ Box and Tube Furnace Controllers
Program segments into Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers for increased temperature accuracy.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
CC58114PBC-11-zone Controller, Dig, Prog, OTCLindberg 1200C Tube Furnace
CC584343PC3-zone Controller, Dig, ProgLindberg 1200C Tube Furnace
CC58114PBA-1Split Hinge Tube Furnace Control OTCLindberg 1200C Split-Hinge Furnace
CC584343PBC-13-zone Controller, Prog, OTCLindberg 1200C Tube Furnace
CC58434BC-13-zone Controller, Dig, Prog, OTCLindberg 1200C Tube Furnace
CC58114A-1Single-zone Controller, DigitalLindberg 1200C Tube Furnace
Catalog number CC58114PBC-1
Price (INR)
-
Request A Quote
Type:
1-zone Controller, Dig, Prog, OTC
For Use With (Equipment):
Lindberg 1200C Tube Furnace
Program segments into Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers for increased temperature accuracy. Features overtemperature control and multiple segment configuration; designed to work with the 1200°C split-hinge and 1500°C box furnaces.
Features
  • Fully wired with advanced microprocessor-based digital control, solid-state power module, on/off curcuit breaker and thermocouple input jacks for each zone
  • Built-in adjustable high-limit overtemperature protection
  • LED simultaneously displays actual temperature vs. setpoint in °C or °F
  • Designed for operation on 120, 208, or 240V 50/60Hz, single-phase line
Overtemperature Control
  • Adjustable digital control is factory installed
  • Protects furnace and load in the event of primary control curcuit failure
  • Overrides main controller and shuts off power to furnace if high limit is reached
  • Must be manually reset for safety
  • Operates via magnetic contacts through a signal from an independent thermocouple
1200°C Three Zone, 16 Segment, One Programmable Controller for Split-hinge Furnaces (CC58434C)
  • Three microprocessor-based PID controls, single segment, single setpoint, one ramp to set point
1200°C Three Zone, 16 Segment, Three Programmable Controllers for Split-hinge Furnaces (CC58434PC)
  • Center Zone—One microprocessor-based PID programmable control, with one program of 16 segments for ramps (up and down) and eight dwells (timed holds)
  • End Zones—Two microprocessor-based PID control, single segment, single setpoint; microprocessers mimic the programmed profile of the center zone controller, but allow a digitally displayed offset up to 100°C
1200°C Digital, Single Setpoint Controllers for Split-hinge Furnaces (CC58114PBA, CC58114PA)
  • Includes one microprocessor-based PID control, single segment, single setpoint, one ramp to setpoint
  • CC58114PBA includes overtemperature control
1500°C Digital, Single Setpoint Controller for Box Furnaces (CC58125C)
  • Includes three microprocessor-based PID controls, single segment, single setpoint, one ramp to set point
1500°C Digital, Single Program, Multiple Segment Programmable Controllers for Box Furnaces (CC58125PC, CC58125PBC)
  • Advanced microprocessor-based PID control, single program with multiple segments for ramp and dwell temperature control
  • CC58125PBC includes overtemperature control
Specifications
Amperage30 A
DisplayDigital
Frequency50/60 Hz
For Use With (Equipment)Lindberg 1200C Tube Furnace
Temperature (Metric)1200°C (Split-Hinge), 1500°C (Box Furnaces)
Type1-zone Controller, Dig, Prog, OTC
Voltage208/240 V
Unit SizeEach