CD19: According to the literature this antibody recognizes the CD19 B cell differentiation antigen. This antigen is expressed on BRead more
Have Questions?
Catalog Number
Quantity
CD19AMA
also known as CD19/A/M-A
0.75 mL
Catalog number CD19AMA
also known as CD19/A/M-A
Price (INR)
-
Quantity:
0.75 mL
CD19: According to the literature this antibody recognizes the CD19 B cell differentiation antigen. This antigen is expressed on B cells throughout their development but is not expressed on terminally differentiated plasma cells. CD19 acts as a coreceptor for B cell activation.α: These fragments recognize B-cells expressing IgA.μ: These fragments recognize B-cells expressing IgM.
Workshop: SJ25-C1: Leukocyte Typing II and IIIα/μ: N/A
Isotype: MG106 and 10305
Class: Class I (ASR)
Statement:
ASR: Analyte Specific Reagent. Analytical and performance characteristics are not established.