MAS™ chemTRAK™ H General Chemistry Controls
MAS™ chemTRAK™ H General Chemistry Controls
Actual product may vary
Thermo Scientific™
MAS™ chemTRAK™ H General Chemistry Controls
Ensure accurate results in general chemistry and TDM methods with comprehensive, multi-analyte Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLevel
CHU-1chemTRAK H Control, Unassayed1
CHA-1chemTRAK H Control, Assayed1
CHA-2chemTRAK H Control, Assayed2
CHU-2chemTRAK H Control, Unassayed2
CHU-3chemTRAK H Control, Unassayed3
CHA-3chemTRAK H Control, Assayed3
Catalog number CHU-1
Price (INR)
-
Request A Quote
Description:
chemTRAK H Control, Unassayed
Level:
1
Ensure accurate results in general chemistry and TDM methods with comprehensive, multi-analyte Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls. These liquid, ready-to-use controls are available in a convenient Vista™ vial format in both assayed and unassayed formats.
  • Human serum matrix
  • Assayed values available for most major analyzers
  • Refer to package insert for specific analyte and stability claims
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionchemTRAK H Control, Unassayed
Level1
Packaging10 Pk.
Quantity15 mL
TypeControl
Detectable AnalytesAcetaminophen, ACP, ALT, Albumin, Alkaline Phosphatase, Amikacin, Amylase, Apolipoprotein A1, Apolipoprotein B, AST, CO2, Bilirubin Direct (DBIL), Bilirubin Total (TBIL), BUN, C3 Complement, C4 Complement, Caffeine, Calcium, Carbamazepine, Chloride, Cholesterol, LDL, HDL, Cholinesterase, Creatine Kinase (CK), Creatinine, Lipoprotein, Digoxin, Disopyramide, Ethanol, Ethosuximide, Ferritin, GGT, Gentamicin, Glucose, Glutamate Dehydrogenase (GLDH), Haptoglobin, HBDH, IgA, IgG, IgM, Iron, TIBC, UIBC, LDH, Lactic Acid, Lidocaine, Lipase, Lithium, Magnesium, Methotrexate, N-acetylprocainamide (NAPA), Osmolality, Phenobarbital, Phenytoin, Phosphorus, Potassium, Prealbumin, Primidone, Procainamide, Quinidine, Salicylate (SALY), Sodium, T3, T4, Theophylline, hTSH, Tobramycin, Total Protein, Transferrin (TRF), Tricyclic Antidepressants, Triglycerides, T-uptake, Uric Acid, Valproic Acid, Vancomycin
Sample TypeSerum
Unit SizeEach
Contents & Storage
30 month shelf-life at -25° to -15°C; 14 day open vial stability at 2° to 8°C unassayed; 7 day open-vial stability at 2° to 8°C assayed; 60 day unopened-vial stability at 2° to 8°C