Cindion™ Combustion Ion Chromatography System
Cindion™ Combustion Ion Chromatography System
Cindion™ Combustion Ion Chromatography System
Thermo Scientific™
Cindion™ Combustion Ion Chromatography System
Perform analysis of total halogens and sulfur in difficult-to-analyze samples with the Thermo Scientific™ Cindion™ Combustion Ion Chromatography (C-IC) System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
CINDION-03Combustion IC for Liquid Samples
CINDION-01Combustion IC for Solid Samples
CINDION-02C-IC for Solid Samples, Aqueous Sample Adsorption (AOX, AOF)
CINDION-04Combustion IC for Gas Samples
Catalog number CINDION-03
Price (INR)
-
For Use With (Application):
Combustion IC for Liquid Samples
Screen for corrosive and toxic halogens and sulfur compounds in a variety of sample types with the Cindion C-IC system. Whether you are analyzing solid, liquid, or gas samples, the Cindion C-IC system provides automated workflows with robust performance and dependable reliability. Experience a seamless, end-to-end C-IC workflow from this single-source combustion IC system.
Small System Footprint
  • Novel, compact combustion tube design ensures complete combustion in a shorter distance
  • Autosampler and absorption tube are integrated into the combustion/absorption module for additional space-savings
Designed for PFAS Analysis
  • Incorporates non-detectable PFAS-containing components within the sample pathway
  • Provides low background levels and highly sensitive detection for PFAS screening
  • Perform PFAS screening following US EPA Method 1621
Single-Source Solution
Acquire the complete system, consumables, and software from a single vendor for a seamless customer experience.
Multiple Configurations for Application Flexibility
  • Different system configurations to accommodate solid, liquid, and gas samples
  • Specific system configuration to measure adsorbable organic fluorine (AOF) by combustion IC
Specifications
For Use With (Application)Combustion IC for Liquid Samples
IncludesCindion Combustion/Absorption Module, Cindion Liquids Kit, Cindion Autosampler, Dionex Inuvion IC System with RFIC
ModelCindion C-IC System for Liquid Sample Analysis
TypeCombustion IC System
Unit SizeEach
Frequently asked questions (FAQs)
Can the Cindion C-IC system be used for US EPA Method 1621 (Adsorbable Organic Fluorine (AOF) by C-IC)?
Yes. The Cindion C-IC system supports US EPA Method 1621-compliant AOF analysis using a fully PFAS-free workflow that features an adsorption module that can process up to six samples in parallel using granular activated carbon (GAC) columns.
What software program(s) are required to run the Cindion C-IC system?
Thermo Scientific Chromeleon Chromatography Data System (CDS) is the only software needed to run the Cindion C-IC system and analyze the results.
Can the IC system used with the Cindion C-IC system be used in stand-alone mode while also being connected to the combustion/absorption module?
Yes. The IC system can be configured with an auxiliary valve that enables you to switch sample delivery between the Cindion Combustion/Absorption module (for combustion IC analysis) and a Thermo Scientific Dionex AS-AP Autosampler (for standalone IC analysis). This process is automated using the Thermo Scientific Chromeleon Chromatography Data System (CDS).
What ion chromatography systems are supported for use with the Cindion C-IC system?
The Thermo Scientific Dionex Inuvion and Thermo Scientific Dionex Integrion IC systems are the only systems that have been tested as part of the Cindion C-IC system.
How has the Cindion C-IC system achieved a relatively small footprint?
By combining the combustion oven, absorption module, and autosampler into one integrated instrument, the Cindion C-IC system minimizes bench space requirements. The compact Z-fold combustion tube also helps by offering efficient combustion within a smaller oven.