MacConkey Agar (Dehydrated)
Actual product may vary
Thermo Scientific™
MacConkey Agar (Dehydrated)
Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with this chromogenic medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0007R2.5kg
CM0007B500 g
Catalog number CM0007R
Price (INR)
-
Quantity:
2.5kg
Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with Thermo Scientific™ Oxoid™ MacConkey Agar (Dehydrated). This differential medium gives a number of diagnostic indications such as bile tolerance, colony morphology and chromogenesis. MacConkey Agar corresponds to the medium recommended by the World Health Organizaton1, the Department of Health2 for the bacterial examination of water.
Use MacConkey Agar for the cultivation of pathogens which may be present in a variety of specimens.
  • Easy-to-read: easy to differentiate Escherichia coli (red non-mucoid colonies) from Staphylococcus spp. (pale pink opaque colonies) and Pseudomonas aeruginosa (green-brown) colonies.
  • Recommended by World Health Organizaton1, the Department of Health2.
Due to its ability to support the growth of pathogenic Gram-positive cocci (e.g. staphylococci and enterococci) as well as Enterobacteriaceae, MacConkey Agar is particularly recommended for the cultivation of pathogens which may be present in a variety of specimens such as urine, feces and wound swabs. While it is selective it does not suppress a mixed bacterial flora to the same extent as other inhibitory media (including other MacConkey agars). MacConkey Agar should be used in parallel with other selective indicator media such as Desoxycholate Citrate Agar, Bismuth Sulphite Agar, Brilliant Green Agar and Brilliant Green Bile (2%) Broth, and a non-selective medium such as Blood Agar.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
General References:
  1. World Health Organization (1963) International Standards for Drinking Water 2nd ed. WHO, Geneva.
  2. Environment Agency (2002) The Microbiology of Drinking Water 2002. Methods for examination of Waters and Associated Materials
Specifications
DescriptionMacConkey Agar
Quantity2.5kg
YieldFor 9.6 L mediμm
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach