Lab-Lemco Nutrient Broth (Dehydrated)
Thermo Scientific™
Lab-Lemco Nutrient Broth (Dehydrated)
Examine water, sewage, and dairy products by American standard methods using this nutritious medium.
Have Questions?
Catalog NumberQuantity
CM0015B500 g
Catalog number CM0015B
Price (INR)
-
Quantity:
500 g
Examine water, sewage, and dairy products by American standard methods using Thermo Scientific™ Oxoid™ Lab-Lemaco Nutrient Broth (Dehydrated). It is used as a nutrient meat extract broth for general bacteriology.
Lab-Lemco Broth is a general purpose liquid medium used for the examination of water, dairy and food products1,2,3
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
General References:
  1. American Public Health Association (1980) Standard Methods for the Examination of Water and Wastewater. 15th Edn. APHA Inc. Washington DC.
  2. American Public Health Association (1978) Standard Methods for the Examination of Dairy Products. 14th Edn. APHA Inc. Washington DC.
  3. American Public Health Association (2001) Compendium of Methods for the Microbiological Examination of Foods 4th Ed
Specifications
DescriptionLab-Lemco Broth (Nutrient Agar) (US formulation)
Quantity500 g
YieldFor 62.5L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach