Lab-Lemco Agar (Dehydrated)
Thermo Scientific™
Lab-Lemco Agar (Dehydrated)
Examine water and dairy products with this nutritious, general purpose medium.
Have Questions?
Catalog NumberQuantity
CM0017B500 g
Catalog number CM0017B
Price (INR)
-
Quantity:
500 g
Examine water and dairy products with nutritious Thermo Scientific™ Oxoid™ Lab-Lemco Agar (Dehydrated)1,2. It is a general-purpose medium for general bacteriological use, and may also be used as the basis for selective, differential, or enriched media. The absence of sodium chloride in the formulation prevents Proteus mirabilis from swarming colonies.
Use Lab-Lamco Agar as a general purpose medium for bacteriological purposes.
It can also be used as nutrient meat extract agar for general bacteriology and for the preservation of stock cultures.
Not all products are available for sale in all territories. Please inquire. 
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
General References:
  1. American Public Health Association (1980) Standard Methods for the Examination of Water and Wastewater. 15th Edn. APHA Inc. Washington DC.
  2. American Public Health Association (1978) Standard Methods for the Examination of Dairy Products. 14th Edn. APHA Inc. Washington DC
Specifications
DescriptionLab-Lemco Agar (Nutrient Agar, U.S. Formulation)
Quantity500 g
YieldFor 21.7L medium
FormPowder
Product TypeAgar
Unit SizeEach