Lactose Broth (Dehydrated)
Thermo Scientific™
Lactose Broth (Dehydrated)
Presumptively identify coliform organisms in milk, water and foods with this medium as specified by the American Public Health Association.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0137B500 g
CM0137R2.5 kg
CM0137T5kg
Catalog number CM0137B
Price (INR)
-
Quantity:
500 g
Presumptively identify coliform organisms in milk, water and foods as specified by the American Public Health Association1,2,3 with Thermo Scientific™ Oxoid™ Lactose Broth (Dehydrated). Beef extracts and peptone in this liquid medium supply essential nutrients while lactose is a fermentable carbohydrate for coliforms. Coliforms ferment lactose in the medium resulting in gas formation which is indicative of positive results.
Tubes of Lactose Broth are inoculated with dilutions of the samples and incubated at 35°C. Examination for gas formation is carried out after 24 and 48 hours incubation. This presumptive evidence of coliform organisms must be confirmed by further tests.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
General References:
  1. American Public Health Association (1978) Standard Methods for the Examination of Dairy Products. 14th Edn. APHA Washington DC.
  2. American Public Health Association (1980) Standard Methods for the Examination of Water and Wastewater. 15th Edn. APHA Inc. Washington DC.
  3. American Public Health Association (1998) Compendium of Methods for the Microbiological Examination of Foods 3rd Edition. APHA Inc. Washington DC.
Specifications
Certifications/ComplianceIndustrial Reference: AFNOR, AOAC, BAM, COMPF, EPA, FDA, USDA, USP.
DescriptionLactose Broth
Quantity500 g
YieldFor 38.5L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach