Reinforced Clostridial Agar (RC Agar) (Dehydrated)
Thermo Scientific™

Reinforced Clostridial Agar (RC Agar) (Dehydrated)

Enumerate and cultivate clostridia and other anaerobes occurring in food and pathological samples with this solid medium.
Have Questions?
Catalog NumberFormQuantity
CM0151BPowder500 g
Catalog number CM0151B
Price (INR)
-
Form:
Powder
Quantity:
500 g
Cultivate and enumerate anaerobes, especially Clostridium species, with Thermo Scientific™ Oxoid™ Reinforced Clostridial Agar (RC Agar) (Dehydrated). A solid, highly nutritious medium, it can be used as an enrichment medium to optimize the growth of clostridia and other anaerobes, lactobacilli, and many other species of bacteria.

Use Reinforced Clostridial Agar for the isolation and cultivation of anaerobic organisms occurring in a variety of habitats, including food and pathological specimens.

Reinforced Clostridial Agar is a solid version of Thermo Scientific Oxoid Reinforced Clostridium Medium suitable for the cultivation and enumeration of clostridia and other anaerobes. Reinforced Clostridial Agar does not differ significantly in performance from the pork-starch-pea agar of Anderson1for the count of anaerobes.

It is employed for the estimation of clostridia in food—see below (also Barnes et al.2 and Angelotti3). Attenborough and Scarr4employed RC Agar, in conjunction with membrane filters, for the count of Clostridium thermosaccharolyticum in sugar.

Reinforced Clostridial Agar is also frequently employed for the investigation of intestinal flora: Perry et al.5for investigation of bovine rumen streptococci; Williams Smith & Crabb6 used the Oxoid medium with added sodium chloride or blood for counts on human or animal feces; Barnes & Goldberg7 employed the Oxoid medium with added chlortetracycline hydrochloride or sodium azide and ethyl violet, for the examination of poultry feces; the Oxoid medium was also used by Goldberg et al.8 for the examination of poultry fecal samples; Williams Smith9 employed Oxoid Reinforced Clostridial Agar with added blood for the ‘total’ and Lactobacillus count of human and animal feces; with added blood and neomycin for determination of Bacteroides, anaerobic Gram-negative cocci, ‘total’ streptococci, sporeformers, yeasts, and ‘actino’ types.

Sneath10 used Oxoid RC Agar and other media for the anaerobic count of microorganisms from soil samples up to approximately 300 years old. Gregory et al.11 also employed the Oxoid medium for the estimation of anaerobes in molding hay.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  

General References:

  1. Anderson A. A. (1951) J. Bact. 62. 425-430.
  2. Barnes Ella M., Despaul J. E. and Ingram M. (1963) J. Appl. Bact. 26. 415-427.
  3. Angelotti R., Hall H. E., Foter M. J. and Lewis K. H. (1962) Appl. Microbiol. 10. 193-199.
  4. Attenborough Sheila J. and Scarr M. Pamela (1957) J. Appl. Bact. 20. 460-466.
  5. Perry K. D., Wilson M. K., Newland L. G. M. and Briggs C. A. E. (1955) J. Appl. Bact. 18. 436-442.
  6. Williams Smith H. and Crabb W. E. (1961) J. Path. Bact. 82. 53-66.
  7. Barnes Ella M. and Goldberg H. S. (1962) J. Appl. Bact. 25. 94-106.
  8. Goldberg H. S., Barnes Ella M. and Charles A. B. (1964) J. Bact. 87. 737-742.
  9. Williams Smith H. (1961) J. Appl. Bact. 24. 235-241.
  10. Sneath P. H. A. (1962) Nature 195. 643-646.
  11. Gregory P. H., Lacey M. E., Festenstein G. N. and Skinner F. A. (1963) J. Gen. Microbiol. 33. 147-174.
Specifications
DescriptionReinforced Clostridial Agar (RC Agar) (Dehydrated)
Media TypeReinforced Clostridial Agar
Quantity500 g
Shelf Life5 Years
FormPowder
Product TypeAgar
Unit SizeEach
Contents & Storage
10°C to 30°C