Clausen Medium-Dithionite Thioglycollate (HS-T) Medium (Dehydrated)
Thermo Scientific™
Clausen Medium-Dithionite Thioglycollate (HS-T) Medium (Dehydrated)
Confidently perform sterility testing using this medium developed by Oslo University.
Have Questions?
Catalog number CM0353B
Price (INR)
-
Confidently perform sterility testing using Thermo Scientific™ Oxoid™ Clausen Medium-Dithionite Thioglycollate (HS-T) Medium (Dehydrated). Dithionite-thioglycollate (HS-T) Broth was developed by Clausen in Oslo University and has been recommended for sterility testing by the Nordic Pharmacopoeia Board.
Perform sterility testing with this Nordic Pharmacopoeia Board recommended medium, containing neutralizing compounds and supplementary minerals.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Certifications/ComplianceIndustrial Reference: Nordic Pharmacopeia.
DescriptionClausen Medium-Dithionite Thioglycollate (HS-T) Medium
Quantity500 g
YieldFor 12.5L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach