SIM Medium (Dehydrated)
Thermo Scientific™
SIM Medium (Dehydrated)
Differentiate enteric bacteria on the basis of sulphide production, indole production and motility with this medium.
Have Questions?
Catalog NumberQuantity
CM0435B500 g
Catalog number CM0435B
Price (INR)
-
Quantity:
500 g
Differentiate enteric bacteria on the basis of sulphide production, indole production and motility with Thermo Scientific™ Oxoid™ SIM Medium (Dehydrated). The medium contains sodium thiosulphate and ferrous ammonium sulphate, which together act as indicators for the production of hydrogen sulphide. Tryptone incorporated into the medium is a tryptophan-rich peptone and is degraded by specific bacteria to produce indole. The use of 0.35% agar in the medium results in the production of a semi-solid medium, ideal for the examination of motility.
A motility-indole medium has been found to be helpful in the identification of the Enterobacteriaceae; e.g. in the differentiation of Klebsiella from Enterobacter and Serratia species1. For convenience, these two important tests have been combined with sulphide-production in one tube. The production of hydrogen sulphide is a useful diagnostic test in the identification of enteric bacteria and is helpful in the differentiation between Salmonella and Shigella.
Typical reactions of organisms in SIM medium:
OrganismMotilityH2SIndole
Escherichia coli ATCC® 25922V-+
Proteus hauseri ATCC® 13315+++
Shigella sonnei ATCC® 25931---
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
1. Blazevic D. J. (1968) Appl. Microbiol. 16. 668.
Specifications
Certifications/ComplianceIndustrial Reference: FDA.
DescriptionSIM Medium
Quantity500 g
YieldFor 16.7L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach