Oxoid™ Standard Plate Count Agar (APHA) (Dehydrated)
Thermo Scientific™
Oxoid™ Standard Plate Count Agar (APHA) (Dehydrated)
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this Dehydrated Standard Plate Count Agar (APHA)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0463B500 g
CM0463R2.5 kg
CM0463T5 kg
Catalog number CM0463B
Price (INR)
-
Quantity:
500 g
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
  • Meets formulation of APHA, AOAC and PHLS
  • Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014
For laboratory use only
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceAPHA, AOAC, PHLS, ISO 11133:2014
Quantity500 g
YieldFor 21.3L medium
FormPowder
Product TypeAgar
Test TypeStandard Plate Count for Microorganism
Unit SizeEach