Oxoid™ Standard Plate Count Agar (APHA) (Dehydrated)
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this Dehydrated Standard Plate Count Agar (APHA)
Have Questions?
Change view
Catalog Number
Quantity
CM0463R
2.5 kg
CM0463B
500 g
CM0463T
5 kg
3 Options
Catalog number CM0463R
Price (INR)
-
Quantity:
2.5 kg
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
Meets formulation of APHA, AOAC and PHLS
Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014
For laboratory use only
Specifications
Certifications/ComplianceAPHA, AOAC, PHLS, ISO 11133:2014