Schaedler Anaerobe Broth (Dehydrated)
Thermo Scientific™
Schaedler Anaerobe Broth (Dehydrated)
Grow and isolate aerobic and anaerobic organisms of the intestinal tract and/or carry out MIC studies with this thioglycollate free medium.
Have Questions?
Catalog NumberQuantity
CM0497B500 g
Catalog number CM0497B
Price (INR)
-
Quantity:
500 g
Grow and isolate aerobic and anaerobic organisms of the intestinal tract and/ or carry out minimum inhibitory concentration (MIC) studies of these organisms using Thermo Scientific™ Oxoid™ Schaedler Anaerobe Broth (Dehydrated).
Schandler Anaerobe Broth is free from thioglycollate and is used to create selective conditions under which delicate and more nutritionally exacting microorganisms of the intestinal tract develop, despite the presence of antagonistic organisms. Normally such fastidious microorganisms would be swamped by the growth of enterococci, coliform, bacilli, and other Gram-negative bacilli.
  • Selective against non-target Gram-negative bacteria, especially Escherichia coli
  • Can also be used to determine MIC levels of anaerobic organisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSchaedler Anaerobe Broth
Quantity500 g
YieldFor 18.9L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach